2-Furaldehyde, 98%
2-Furaldehyde, 98%
2-Furaldehyde, 98%
Thermo Scientific Chemicals

2-Furaldehyde, 98%

CAS: 98-01-1 | C5H4O2 | 96.09 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
1000 g
Catalog number A16167.0B
also known as A16167-0B
Price (USD)
63.70
Each
Quantity:
1000 g
Request bulk or custom format
Price (USD)
63.70
Each
Chemical Identifiers
CAS98-01-1
IUPAC Namefuran-2-carbaldehyde
Molecular FormulaC5H4O2
InChI KeyHYBBIBNJHNGZAN-UHFFFAOYSA-N
SMILESO=CC1=CC=CO1
View more
SpecificationsSpecification SheetSpecification Sheet
FormLiquid
Appearance (Color)Clear pale yellow to orange to brown to dark brown
Assay (GC)≥97.5% (non-U.S. specification)
Assay from Suppliers CofA≥97.5% (U.S. specification)
Identification (FTIR)Conforms (non-U.S. specification)
View more
2-Furaldehyde is used in the manufacture of furfural-phenol plastics such as Durite; in solvent refining of petroleum oils and in the preparation of pyromucic acid. It is used as a solvent for nitrated cotton, cellulose acetate and gums; for accelerating vulcanization; in the synthesis of furan derivatives.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Furaldehyde is used in the manufacture of furfural-phenol plastics such as Durite; in solvent refining of petroleum oils and in the preparation of pyromucic acid. It is used as a solvent for nitrated cotton, cellulose acetate and gums; for accelerating vulcanization; in the synthesis of furan derivatives.

Solubility
Soluble in water (83 g/L).

Notes
Air sensitive. Incompatible with oxidizing agents and acids.
RUO – Research Use Only

General References:

  1. Michael Jerry Antal Jr.; Tongchit Leesomboon; William S.Mok; Geoffrey N.Richards. Mechanism of formation of 2-furaldehyde from d-xylose. Carbohydrate Research. 1991, 217, 71-85
  2. Michael S.McClure; Bobby Glover; Ellen McSorley; Alan Millar; Martin H.Osterhout; Frank Roschangar. Regioselective palladium-catalyzed arylation of 2-furaldehyde. Org. Lett. 2001, 3, (11),1677-1680