2-Acetyl-5-methylfuran, 98+%
2-Acetyl-5-methylfuran, 98+%
2-Acetyl-5-methylfuran, 98+%
Thermo Scientific Chemicals

2-Acetyl-5-methylfuran, 98+%

CAS: 1193-79-9 | C7H8O2 | 124.139 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
25 g
50 g
Catalog number A16172.09
also known as A16172-09
Price (USD)
28.65
Special offer
Online exclusive
Ends: 30-Jun-2026
34.00
Save 5.35 (16%)
Each
Quantity:
10 g
Request bulk or custom format
Price (USD)
28.65
Special offer
Online exclusive
Ends: 30-Jun-2026
34.00
Save 5.35 (16%)
Each
Chemical Identifiers
CAS1193-79-9
IUPAC Name1-(5-methylfuran-2-yl)ethan-1-one
Molecular FormulaC7H8O2
InChI KeyKEFJLCGVTHRGAH-UHFFFAOYSA-N
SMILESCC(=O)C1=CC=C(C)O1
View more
SpecificationsSpecification SheetSpecification Sheet
Identification (FTIR)Conforms
Refractive Index1.5115-1.5155 @ 20?C
FormLiquid
Appearance (Color)Clear yellow
Assay (GC)≥98.0%

Glucose was transformed to various furan compounds and carboxylic acids derivatives by heat treatment of autoclave such as levulinic acid, 2-acetylfuran and 2-acetyl-5-methylfuran. using 2-acetyl-5-methylfuran or 1,5-dimethyl-2-pyrrolecarbonitrile as reactants, that the formation of the 4-arylation products. Reaction of 2-acetyl-5-methylfuran 6a with 2-aminoethanol pioduced in 79% yield the aminal.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Glucose was transformed to various furan compounds and carboxylic acids derivatives by heat treatment of autoclave such as levulinic acid, 2-acetylfuran and 2-acetyl-5-methylfuran. using 2-acetyl-5-methylfuran or 1,5-dimethyl-2-pyrrolecarbonitrile as reactants, that the formation of the 4-arylation products. Reaction of 2-acetyl-5-methylfuran 6a with 2-aminoethanol pioduced in 79% yield the aminal.

Solubility
Slightly soluble in water. Soluble in alcohol.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
RUO – Research Use Only

General References:

  1. Keith Basey,; Jack G. Woolley. Alkaloids of Physalis alkekengi. Phytochemistry. 1973, 12 (10), 2557-2559.
  2. Liang Zhang,; Isao Kuniyoshi,; Mitsuyo Hirai,; Makoto Shoda. Oxidation of dimethyl sulfide byPseudomonas acidovorans DMR-11 isolated from peat biofilter. Biotechnology Letters. 1991, 13 (3), 223-228.