Who can I contact for questions regarding the hPSC Scorecard Assay Service (Cat. No. A16179)?
CUB Characterization Services can be contacted at the following Email. However, please keep in mind that stem cell characterization services were discontinued effective Feb 27th, 2025, so we are no longer able to provide this service.