(-)-Dibenzoyl-L-tartaric acid,98%
(-)-Dibenzoyl-L-tartaric acid,98%
(-)-Dibenzoyl-L-tartaric acid,98%
Thermo Scientific Chemicals

(-)-Dibenzoyl-L-tartaric acid,98%

CAS: 2743-38-6 | C18H14O8 | 358.30 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
500 g
Catalog number A16181.36
also known as A16181-36
Price (USD)
225.65
Online Exclusive
251.00
Save 25.35 (10%)
Each
Quantity:
500 g
Request bulk or custom format
Price (USD)
225.65
Online Exclusive
251.00
Save 25.35 (10%)
Each
Chemical Identifiers
CAS2743-38-6
IUPAC Name(2R,3R)-2,3-bis(benzoyloxy)butanedioic acid
Molecular FormulaC18H14O8
InChI KeyYONLFQNRGZXBBF-ZIAGYGMSSA-N
SMILESOC(=O)[C@H](OC(=O)C1=CC=CC=C1)[C@@H](OC(=O)C1=CC=CC=C1)C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to pale cream
Assay (Aqueous acid-base Titration)≥97.5 to ≤102.5%
Optical Rotation-120? ? 3? (c=5 in methanol)
FormPowder or crystalline powder
(-)-Dibenzoyl-L-tartaric acid, anhydrous is used as a reagent used to produce chiral salts. Also used as organic chemical synthesis intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(-)-Dibenzoyl-L-tartaric acid, anhydrous is used as a reagent used to produce chiral salts. Also used as organic chemical synthesis intermediate.

Solubility
Slightly soluble in water.

Notes
It is sensitive to moisture. Incompatible with oxidizing agents.
RUO – Research Use Only
Malachy McCann; Fergal Humphreys; Vickie McKee. Transition metal complexes of dibenzoyl-L-tartaric acid (db-L-tarH2) and L-tartaric acid (L-tarH2) ; X-ray crystal structure of {[Cu(L-tar) (phen)] · 6H2O}n (phen = 1,10-phenanthroline). Polyhedron. 1997, 16, (20),3655-3661