2,6-Dichloro-4-fluorophenol, 98%
2,6-Dichloro-4-fluorophenol, 98%
2,6-Dichloro-4-fluorophenol, 98%
2,6-Dichloro-4-fluorophenol, 98%
Thermo Scientific Chemicals

2,6-Dichloro-4-fluorophenol, 98%

CAS: 392-71-2 | C6H3Cl2FO | 180.987 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
100 g
Catalog number A16193.22
also known as A16193-22
Price (USD)
558.65
Online Exclusive
620.00
Save 61.35 (10%)
Each
Add to cart
Quantity:
100 g
Request bulk or custom format
Price (USD)
558.65
Online Exclusive
620.00
Save 61.35 (10%)
Each
Add to cart
Chemical Identifiers
CAS392-71-2
IUPAC Name2,6-dichloro-4-fluorophenol
Molecular FormulaC6H3Cl2FO
InChI KeyBOJVIFKSTRCIRJ-UHFFFAOYSA-N
SMILESOC1=C(Cl)C=C(F)C=C1Cl
View more
SpecificationsSpecification SheetSpecification Sheet
Melting Point (clear melt)46.0-52.0?C
Assay (GC)≥97.5%
Appearance (Color)Cream to orange to pale brown
FormCrystals or powder or crystalline powder or lumps or fused solid
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only