Benzo-18-crown-6, 97%
Benzo-18-crown-6, 97%
Benzo-18-crown-6, 97%
Thermo Scientific Chemicals

Benzo-18-crown-6, 97%

CAS: 14098-24-9 | C16H24O6 | 312.362 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
25 g
Catalog number A16223.06
also known as A16223-06
Price (USD)
173.65
Online Exclusive
193.00
Save 19.35 (10%)
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
173.65
Online Exclusive
193.00
Save 19.35 (10%)
Each
Chemical Identifiers
CAS14098-24-9
IUPAC Name2,3,5,6,8,9,11,12,14,15-decahydro-1,4,7,10,13,16-benzohexaoxacyclooctadecine
Molecular FormulaC16H24O6
InChI KeyDSFHXKRFDFROER-UHFFFAOYSA-N
SMILESC1COCCOCCOC2=CC=CC=C2OCCOCCO1
View more
SpecificationsSpecification SheetSpecification Sheet
FormCrystals or powder or crystalline powder
Identification (FTIR)Conforms
Melting Point (clear melt)39.0-48.0?C
Appearance (Color)White to pale yellow
Assay (GC)≥96.0%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only