4-(Methylamino)phenol sulfate, 99%
4-(Methylamino)phenol sulfate, 99%
4-(Methylamino)phenol sulfate, 99%
Thermo Scientific Chemicals

4-(Methylamino)phenol sulfate, 99%

CAS: 55-55-0 | C14H18N2O2·H2SO4
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
250 g
1000 g
Catalog number A16236.18
also known as A16236-18
Price (USD)
27.65
Online Exclusive
30.40
Save 2.75 (9%)
Each
Quantity:
50 g
Request bulk or custom format
Price (USD)
27.65
Online Exclusive
30.40
Save 2.75 (9%)
Each
Chemical Identifiers
CAS55-55-0
IUPAC Namebis(4-(methylamino)phenol); sulfuric acid
Molecular FormulaC14H20N2O6S
InChI KeyZVNPWFOVUDMGRP-UHFFFAOYSA-N
SMILESOS(O)(=O)=O.CNC1=CC=C(O)C=C1.CNC1=CC=C(O)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
FormCrystals or powder or crystalline powder
Identification (FTIR)Conforms
Appearance (Color)White to pale cream or pale grey
Assay (Aqueous acid-base Titration)≥98.5% to ≤101.5%

4-(Methylamino)phenol sulfate, is a is a popular photographic developer used in black & white photography. It is also an excellent developing agent for most continuous tone developer applications, and it has been widely used in published developer formulas as well as commercial products.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-(Methylamino)phenol sulfate, is a is a popular photographic developer used in black & white photography. It is also an excellent developing agent for most continuous tone developer applications, and it has been widely used in published developer formulas as well as commercial products.

Solubility
It is soluble in water (50 mg/mL).

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from acids, acid chlorides, acid anhydrides, oxidizing agents.
RUO – Research Use Only

General References:

  1. AR Tiedemann.; TD Anderson. Rapid analysis of total sulphur in soils and plant material. Plant and Soil. 1971, 35, (1), 197-200.
  2. LC Raiford.; GO Inman. Benzoxazolone Formation in the Attempt to Prepare Certain Mixed Diacyl Derivatives of o-Aminophenol. J. Am. Chem. Soc. 1934, 56, (7), 1586-1590.