Magnesium stearate
Magnesium stearate
Magnesium stearate
Thermo Scientific Chemicals

Magnesium stearate

CAS: 557-04-0 | C36H70MgO4 | 591.257 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
1000 g
Catalog number A16244.0B
also known as A16244-0B
Price (USD)
177.00
Each
Quantity:
1000 g
Request bulk or custom format
Price (USD)
177.00
Each
Chemical Identifiers
CAS557-04-0
IUPAC Namemagnesium(2+) dioctadecanoate
Molecular FormulaC36H70MgO4
InChI KeyHQKMJHAJHXVSDF-UHFFFAOYSA-L
SMILES[Mg++].CCCCCCCCCCCCCCCCCC([O-])=O.CCCCCCCCCCCCCCCCCC([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
FormPowder
Assay from Suppliers CofAStearic acid: ≥40.0% (GC)
Impurity contentLead, Pb: ≤10ppm
Impurity contentSulfate, SO4: ≤1%
Assay from Suppliers CofAStearic & Palmitic acid: ≥90.0% (GC)
View more
Magnesium stearate is involved in the production of medical tablets, capsules, powders and polymer formulation. It is also used as a lubricant for tablets, anti-adherent, in dry coating and as a binding agent. It is an important ingredient in baby formulas. Further, it is used in the hydrogenation process. In addition to this, it is used to bind the sugar in hard candies like mints.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Magnesium stearate is involved in the production of medical tablets, capsules, powders and polymer formulation. It is also used as a lubricant for tablets, anti-adherent, in dry coating and as a binding agent. It is an important ing
RUO – Research Use Only

General References:

  1. Lebeau, D.; Beuvier, L.; Cornaton, M.; Miserque, F.; Tabarant, M.; Esnouf, S.; Ferry, M. Aging of magnesium stearate under high doses gamma irradiation and oxidative conditions. J. Nucl. Mater. 2015, 460, 130-138.
  2. Haware, R. V.; Shivagari, R.; Johnson, P. R.; Staton, S.; Stagner, W. C.; Gupta, M. R. Application of Multivariate Methods to Evaluate the Functionality of Bovine- and Vegetable-Derived Magnesium Stearate. J. Pharm. Sci. 2014, 103 (5), 1466-1477.