Mercury(I) nitrate dihydrate, 97%
Thermo Scientific Chemicals

Mercury(I) nitrate dihydrate, 97%

CAS: 7782-86-7 | Hg2(NO3)2·2H2O
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
250 g
1000 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number A16268.18
also known as A16268-18
Price (USD)
-
Quantity:
50 g
Request bulk or custom format
Chemical Identifiers
CAS7782-86-7
IUPAC Namedimercury(1+) dihydrate dinitrate
Molecular FormulaH4Hg2N2O8
InChI KeyLSABZDVKJBWCBE-UHFFFAOYSA-N
SMILESO.O.[Hg+].[Hg+].[O-][N+]([O-])=O.[O-][N+]([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
CommentMay contain small amounts of dark particulate matter as part of manufacturing process
Appearance (Color)White to yellow to yellow-red
FormCrystalline or lumpy powder
Assay (unspecified)≥96.0%
Mercury(I) nitrate dihydrate is used in the preparation of other mercury(I) compounds. It acts as a reducing agent. It is involved in the preparation of mercury(II) nitrate by treating with dilute nitric acid as well as to get mercury on exposure to light.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Mercury(I) nitrate dihydrate is used in the preparation of other mercury(I) compounds. It acts as a reducing agent. It is involved in the preparation of mercury(II) nitrate by treating with dilute nitric acid as well as to get mercury on exposure to light.

Solubility
Slightly soluble in water.

Notes
Light sensitive. Incompatible with alcohols, sulfur and strong reducing agents.
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
RUO – Research Use Only

General References:

  1. Laborda, E.; Rogers, E. I.; Martínez-Ortiz, F.; Molina, Á.; Compton, R. G. Application of double pulse theory for hemispherical microelectrodes to the experimental study of slow charge transfer processes. Electrochim. Acta 2014, 55 (22), 6577-6585.
  2. Dehghanpour, S.; Khalaj, M.; Mahmoudi, A. Mercury(I) complexes with bidentate iminopyridine ligands: Synthesis, spectral characterization and structural analysis. Polyhedron 2009, 28 (7), 1205-1210.