1,4-Dichlorobenzene, 99+%
1,4-Dichlorobenzene, 99+%
1,4-Dichlorobenzene, 99+%
Thermo Scientific Chemicals

1,4-Dichlorobenzene, 99+%

CAS: 106-46-7 | C6H4Cl2 | 146.998 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
2500 g
Catalog number A16290.22
also known as A16290-22
Price (USD)
24.65
Online Exclusive
26.70
Save 2.05 (8%)
Each
Quantity:
100 g
Request bulk or custom format
Price (USD)
24.65
Online Exclusive
26.70
Save 2.05 (8%)
Each
Chemical Identifiers
CAS106-46-7
IUPAC Name1,4-dichlorobenzene
Molecular FormulaC6H4Cl2
InChI KeyOCJBOOLMMGQPQU-UHFFFAOYSA-N
SMILESClC1=CC=C(Cl)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
FormCrystals or powder or crystalline powder or lumps
Identification (FTIR)Conforms
Melting Point (clear melt)50.0-57.0?C
Assay (GC)≥99.0%
Appearance (Color)White
1,4-Dichlorobenzene, is used as moth repellant, air fresheners and toilet blocks and general deodorant for garbage and restrom as well as to control fruit borers and ants, may be applied to tobacco seed beds for blue mold control. It is also used as a pharmaceutical intermediate. 1,4-dichlorobenzene is as a used as a minor processing aid in the production of grinding wheels.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,4-Dichlorobenzene, is used as moth repellant, air fresheners and toilet blocks and general deodorant for garbage and restrom as well as to control fruit borers and ants, may be applied to tobacco seed beds for blue mold control. It is also used as a pharmaceutical intermediate. 1,4-dichlorobenzene is as a used as a minor processing aid in the production of grinding wheels.

Solubility
It is insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents, aluminum and its alloys, some plastics.
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only

General References:

  1. M Charbonneau.; J Strasser.; EA Lock.; MJ Turner. Involvement of reversible binding to α 2u-globulin in 1, 4-dichlorobenzene-induced nephrotoxicity. Toxicology and Applied Pharmacology. 1989, 99, (1), 122-132.
  2. RR Jones.; RG Bergman. p-Benzyne. Generation as an intermediate in a thermal isomerization reaction and trapping evidence for the 1, 4-benzenediyl structure. J. Am. Chem. Soc. 1972, 94, (2), 660-661.