3,4-Dichlorophenyl isocyanate, 97%
3,4-Dichlorophenyl isocyanate, 97%
3,4-Dichlorophenyl isocyanate, 97%
Thermo Scientific Chemicals

3,4-Dichlorophenyl isocyanate, 97%

CAS: 102-36-3 | C7H3Cl2NO | 188.007 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number A16298.06
also known as A16298-06
Price (USD)
43.65
Online Exclusive
48.50
Save 4.85 (10%)
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
43.65
Online Exclusive
48.50
Save 4.85 (10%)
Each
Chemical Identifiers
CAS102-36-3
IUPAC Name1,2-dichloro-4-isocyanatobenzene
Molecular FormulaC7H3Cl2NO
InChI KeyMFUVCHZWGSJKEQ-UHFFFAOYSA-N
SMILESClC1=CC=C(C=C1Cl)N=C=O
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (GC)≥96.0%
FormCrystals or fused solid
Appearance (Color)White to pale cream
Identification (FTIR)Conforms
Melting Point (clear melt)40-44?C
Methyl-labeled diuron was prepared by the reaction of 3,4-dichlorophenyl- isocyanate with C 'labeled dimethylamine.Substituted urea derivatives were prepared by reacting 3,4-dichlorophenyl isocyanate with amino acids, dipeptides, histamine. The R- and S-isomers and RS-racemate of MBPU were synthesized by allowing equimolar quantities of 3,4-dichlorophenyl- isocyanate

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Methyl-labeled diuron was prepared by the reaction of 3,4-dichlorophenyl- isocyanate with C ′″labeled dimethylamine.Substituted urea derivatives were prepared by reacting 3,4-dichlorophenyl isocyanate with amino acids, dipeptides, histamine. The R- and S-isomers and RS-racemate of MBPU were synthesized by allowing equimolar quantities of 3,4-dichlorophenyl- isocyanate

Solubility
Hydrolyzes in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, moisture, water, amines, alcohols.
RUO – Research Use Only

General References:

  1. F. S. Tanaka,; B. L. Hoffer,; R. G. Wiena. Photolysis of 3-(3,4-dichlorophenyl)-1, 1 -Dimethylurea (Diuron) in dilute aqueous solution . Toxicological & Environmental Chemistry. 1986, 11 (4) ,261-269.
  2. Donald E. Morelanda. Effects of Optically Active 1-(α-Methylbenzyl)-3-(3,4-dichlorophenyl)urea on Reactions of Mitochondria and Chloroplasts 1. Plant Physiology . 1971, 47 (1), 53-58.