(Methoxycarbonylmethyl)triphenylphosphonium bromide, 98+%
(Methoxycarbonylmethyl)triphenylphosphonium bromide, 98+%
(Methoxycarbonylmethyl)triphenylphosphonium bromide, 98+%
Thermo Scientific Chemicals

(Methoxycarbonylmethyl)triphenylphosphonium bromide, 98+%

CAS: 1779-58-4 | C21H20BrO2P | 415.27 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
500 g
Catalog number A16301.22
also known as A16301-22
Price (USD)
92.65
Special offer
Online exclusive
Ends: 30-Jun-2026
109.00
Save 16.35 (15%)
Each
Quantity:
100 g
Request bulk or custom format
Price (USD)
92.65
Special offer
Online exclusive
Ends: 30-Jun-2026
109.00
Save 16.35 (15%)
Each
Chemical Identifiers
CAS1779-58-4
IUPAC Name(2-methoxy-2-oxoethyl)triphenylphosphanium bromide
Molecular FormulaC21H20BrO2P
InChI KeyVCWBQLMDSMSVRL-UHFFFAOYSA-M
SMILES[Br-].COC(=O)C[P+](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
FormPowder
Appearance (Color)White
Assay (Titration ex Bromide)≥98.0 to ≤102.0%
(Methoxycarbonylmethyl)triphenylphosphonium bromide is a reactant for the synthesis of GSK-3 inhibitors using the Wittig reaction, substituted furanones for antifungal and cytostatic activities, photochromic dithienylethene derivatives, gem-diiodoalkanes containing allylic alcohols and highly substituted cyclopentenes and brazilanes via Au-catalyzed deoxygenative Nazarov cyclization. As reactant for asymmetric hydrogenation of α,β-unsaturated ester-phosphonates, studies of thermal decomposition of phosphonium alkyl ester salts.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(Methoxycarbonylmethyl)triphenylphosphonium bromide is a reactant for the synthesis of GSK-3 inhibitors using the Wittig reaction, substituted furanones for antifungal and cytostatic activities, photochromic dithienylethene derivatives, gem-diiodoalkanes containing allylic alcohols and highly substituted cyclopentenes and brazilanes via Au-catalyzed deoxygenative Nazarov cyclization. As reactant for asymmetric hydrogenation of α,β-unsaturated ester-phosphonates, studies of thermal decomposition of phosphonium alkyl ester salts.

Solubility
Soluble in methanol(almost transparency).

Notes
Hygroscopic. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, water, moisture.
RUO – Research Use Only

General References:

  1. Petr Šenel et. al. Antifungal 3,5-disubstituted furanones: From 5-acyloxymethyl to 5-alkylidene derivatives. Bioorganic & Medicinal Chemistry. 2010, 18 (5), 1988-2000.
  2. Oliver Tosic et. al.Photochromic dithienylethenes with extended pi-systems. Photochemical & Photobiological Sciences. 2010, 9,(2), 128-130.