Potassium bromide, 99+%
Potassium bromide, 99+%
Potassium bromide, 99+%
Thermo Scientific Chemicals

Potassium bromide, 99+%

CAS: 7758-02-3 | BrK | 119.00 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
1000 g
2.5 kg
5000 g
Catalog number A16339.0B
also known as A16339-0B
Price (USD)
129.00
Each
Quantity:
1000 g
Request bulk or custom format
Price (USD)
129.00
Each
Chemical Identifiers
CAS7758-02-3
IUPAC Namepotassium bromide
Molecular FormulaBrK
InChI KeyIOLCXVTUBQKXJR-UHFFFAOYSA-M
SMILES[K+].[Br-]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Assay (Titration ex Bromide)≥99.0 to ≤101.0%
FormCrystals or powder or crystalline powder or lumps or fused solid
Water Content (Karl Fischer Titration)≤1.0%
Used as an analytical reagent and an oxidizing agent. Potassium bromate is used primarily as a maturing agent for flour and as a dough conditioner. It is also used as a laboratory reagent, in permanent-wave compounds such as food additives and explosives.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Used as an analytical reagent and an oxidizing agent. Potassium bromate is used primarily as a maturing agent for flour and as a dough conditioner. It is also used as a laboratory reagent, in permanent-wave compounds such as food additives and explosives.

Solubility
Slightly soluble in alcohol but insoluble in acetone and ethanol.

Notes
Strong oxidizer. Incompatible with combustible materials, organics, reducing agents, aluminum and finely powdered metals
RUO – Research Use Only

General References:

  1. Khan, N.; Sharma, S.; sativa, N. Ameliorates potassium bromate-induced early events of carcinogenesis: diminution of oxidative stress. Hum Exp Toxicol. 2003, 22 (4) , 193-203.
  2. Murata, M.; Bansho, Y.; Inoue, S. ; Ito, K.; Ohnishi, S.; Midorikawa, K.; Kawanishi, S. Requirement of Glutathione and Cysteine in Guanine-Specific Oxidation of DNA by Carcinogenic Potassium Bromate. Chem. Res. Toxicol. 2001, 14 (6), 678-685.