Neural Induction Supplement (50X) - FAQs
Neural Induction Supplement (50X) - FAQs
View additional product information for Neural Induction Supplement (50X) - FAQs (A1647701)
23 product FAQs found
Can PSC Neural Induction Medium be used to differentiate hPSCs into neurospheres or neurons?
PSC Neural Induction Medium is a medium to convert hPSCs into NSCs. At day 7 of neural induction, dissociate P0 NSCs with Accutase. The following recommendations can be use for differentiation of neurons or neurospheres: For differentiation of neurons: Use PSC Neural Induction Medium to convert hPSCs into NSCs. At day 7 of neural induction, dissociate P0 NSCs with Accutase. If dissociated NSCs are plated on a Geltrex-coated culture vessel in neural expansion medium, NSCs will grow as a mono-layer. For NSC expansion, it is necessary to treat NSCs with 5 µM ROCK inhibitor Y27632 at the time of plating to prevent cell death if NSCs are under P4. For differentiation of neurospheres: Use PSC Neural Induction Medium to convert hPSCs into NSCs. At day 7 of neural induction, dissociate P0 NSCs with Accutase. If dissociated NSCs are plated into a non-coated flask in neural expansion medium, dissociated NSCs will re-aggregate into small spheres. NSCs in each sphere will proliferate and form big neurospheres. We have not expanded NSCs in neurosphere format in-house. You can test expanding NSCs in neurospheres with and without ROCK inhibitor Y27632 to determine which is optimal for your workflow. Neural stem cells in adherent or neurosphere conditions can be differentiated into neurons using the appropriate protocol.
Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.
Have you performed flow cytometry or qPCR studies to determine the expression of NKX2.1 in NSCs generated from iPSC using PSC Neural Induction Medium?
Studies have not been performed using flow cytometry or qPCR to detect NKX2.1 expression in NSCs induced by Neural Induction Medium.
Is it normal to see loss of pluripotency markers (i.e. SSEA-4, Oct-4, and Tra1-60) and still test positive for SOX2 expression when NSCs are cultured in neural expansion medium?
Yes, this is normal. SOX2 is expressed by both hPSCs and hPSC-derived NSCs, so you will see its expression in the induced NSCs from iPSCs.
Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.
Can NSCs induced by PSC Neural Induction Medium be differentiated into neurons on glass slides/coverslips coated with laminin and poly-L-ornithine?
NSCs induced using the PSC Neural Induction Medium can be differentiated on glass slides/coverslips, but the glass should be cleaned well before coating. We recommend the following cleaning protocol:
1. Treat glass cover slips with1M HCl at RT for 2-3 h on a shaker.
2. Rinse with tap water 5 times, and then double distilled water for 2 times
3. Store cleaned cover slips in 70% ethanol.
4. Dry cover slips by transferring cover slips into culture plate with sterile forceps before poly-L-ornithine and laminin coating.
Can I maintain differentiated neurospheres in Neurobasal+B27+N2+bFGF+EGF?
Neurospheres can be plated on laminin coated culture plates for neuron differentiation. The issue is that it is difficult to control the plating density of neurospheres. Alternatively, neurospheres can be dissociated into single cells and plate single cell suspension at a certain density such as 1-5 x 10^4 cells/cm2 onto laminin coated plates for neuron differentiation. For general neuron differentiation, Neurobasal+B27+N2 can be used. Growth factors such as BDNF and/or GDNF can be added into medium for improving survival of differentiating NSCs.
Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.
Can I maintain neurospheres in PSC Neural Expansion Medium?
Yes, neurospheres can be maintained in PSC Neural Expansion medium. The recipe for Neural Expansion medium is found in the Neural Induction Medium product manual.
Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.
Why am I seeing abnormal staining pattern of NSCs stained with antibodies against neural markers?
Antibodies that are not stored or handled properly may result in loss of staining quality and should be replaced. Note that Nestin should stain filaments in the cytoplasm whereas Pax6, Sox1, and Sox2 should stain the nucleus. Oct4 should stain the nucleus of pluripotent cells, but not NSCs.
What should I do if I see extensive cell death after thawing and plating cryo-preserved NSCs?
If you are seeing extensive cell death and the NSCs are cryo-preserved under P4, we recommend treating with 5 µM ROCK Inhibitor Y27632 after plating.
Can NSCs induced by PSC Neural Induction Medium be differentiated into motor neurons?
Yes, NSCs induced by PSC Neural Induction Medium can be differentiated into motor neurons. We do not offer a validated protocol, but you can refer to the following publication or perform a literature search for additional methods:
http://rd.springer.com/article/10.1007/s12015-014-9541-0
What PSC cell lines have been tested using the PSC Neural Induction Medium kit?
We have tested the following PSC cell lines for the PSC Neural Induction Medium:
• H9 embryonic stem cells
• iPSC line created in-house
• Episomal iPSC cell line
Can NSCs generated using PSC Neural Induction Medium be expanded in the PSC Neural Induction Medium or do they need to be in the recommended NSC expansion medium, as stated in the product manual?
We recommend using neural expansion medium with the following composition to expand NSCs generated using the PSC Neural Induction Medium: - 50:50 Neurobasal medium: Advanced DMEM/F12 plus PSC Neural Induction Supplement.
PSC lines can behave differently, and we recommend performing your own validation experiments if you want to use PSC Neural Induction Medium, or you do not observe good expansion using the Neurobasal and Advanced DMEM/F12 combination.
Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.
When should I add ROCK inhibitor to PSC Neural Induction Medium?
It is not necessary or recommended to add ROCK inhibitor to PSC Neural Induction Medium when differentiating cells into neural stem cells. You would only add ROCK inhibitor to iPSC or ESC before initiation of neural induction and when expanding neural stem cells using Neural Expansion Medium.
Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.
Can NSCs derived by PSC Neural Induction Medium be differentiated into cells in the peripheral nervous system (PNS)?
No, NSCs derived by PSC Neural Induction Medium cannot be differentiated into cells of the PNS. They can be differentiated into neurons and glial cells in the central nervous system. Sensory, sympathetic and parasympathetic neurons, as well as Schwann cells in the PNS are derived from precursor cells of the neural crest.
Can PSC Neural Induction Medium convert mouse PSCs into NSCs?
PSC Neural Induction Medium has not been tested and is not recommended for the conversion of mouse PSCs into NSCs.
Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.
What should I do if my NSCs reach near maximal confluence within 2-3 days after passaging?
NSCs derived from some human PSC lines may have an increased proliferation rate. In this case, we recommend decreasing NSC plating density to 5x10^4 cells/cm2.
Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.
What should I do if I see extensive cell death after plating dissociated NSCs for expansion and differentiation?
We recommend adding ROCK inhibitor Y27632 into cell suspension to reach a final concentration of 5 µM if the passage number of NSCs is under P4. NSCs from some human PSC lines may be more sensitive to dissociation. For NSCs derived from those PSC lines, the overnight treatment with Y27632 after P4 decreases cell death after re-plating.
Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.
How can I prevent extensive cell death during the late stage of neural induction (Day 5-7) with PSC Neural Induction Medium?
To prevent extensive cell death during late stage neural induction, we recommend checking whether cells are over-confluent. If they are, change the PSC Neural Induction Medium every day with 5 mL per well of a 6-well plate.
Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.
How can I prevent cell detachment during neural induction with PSC Neural Induction Medium?
For PSCs cultured in Essential 8 Medium, cells may detach during neural induction. To prevent cell detachment, we recommend coating your culture plates with Vitronectin at 1µg/cm2 when splitting PSCs.
Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.
How can I remove non-neural cells during NSC expansion following the induction of NSCs from hPSCs using PSC Neural Induction Medium?
We recommend that you discard the culture and begin the process again. You will want to select and maintain a high quality of PSCs before inducing neural differentiation.
Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.
How do I get the right starting density for my human PSCs?
Due to variable parameters such as the confluence of PSCs, cell clump passaging, cell attachment and the property of different PSC lines, it may be difficult to determine the splitting ratio. To obtain the optimal starting density for neural induction with PSC Neural Induction Medium we recommend estimating the cell number of PSC clumps before plating. Instructions for estimating the cell number of PSC clumps can be found in the protocol (https://assets.thermofisher.com/TFS-Assets/LSG/manuals/MAN0008031.pdf). PSC culture should be at 15-25% confluence on day 1 of PSC splitting (Day 0 of neural induction).
Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.
Which format of cultures should I use for neural induction with PSC Neural Induction Medium?
Neural induction can be started with human PSCs cultured on 6-well plate or culture dishes. PSCs cultured in flasks are not recommended because it is difficult to remove non-neural differentiated colonies in flasks.
Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.
Does PSC Neural Induction Medium work for human PSCs cultured on feeders (mouse embryonic fibroblasts, MEF) and feeder-free conditions?
PSC Neural Induction Medium has been tested on human PSCs cultured in both feeder-free and feeder-based conditions. To eliminate MEF contamination, human PSCs cultured in feeder-free conditions such as Essential 8 and StemPro hESC SFM are strongly recommended for starting neural induction.
Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.
Does PSC Neural Induction Medium work for both human ESCs and iPSCs?
Yes, PSC Neural Induction Medium works for both human ESCs and iPSCs.
Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.