(+)-Dibenzoyl-D-tartaric acid monohydrate, 99%
(+)-Dibenzoyl-D-tartaric acid monohydrate, 99%
(+)-Dibenzoyl-D-tartaric acid monohydrate, 99%
Thermo Scientific Chemicals

(+)-Dibenzoyl-D-tartaric acid monohydrate, 99%

CAS: 80822-15-7 | C18H16O9 | 376.32 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Catalog number A16503.22
also known as A16503-22
Price (USD)
129.65
Special offer
Online exclusive
Ends: 30-Jun-2026
152.00
Save 22.35 (15%)
Each
Quantity:
100 g
Request bulk or custom format
Price (USD)
129.65
Special offer
Online exclusive
Ends: 30-Jun-2026
152.00
Save 22.35 (15%)
Each
Chemical Identifiers
CAS80822-15-7
IUPAC Name(2S,3S)-2,3-bis(benzoyloxy)butanedioic acid hydrate
Molecular FormulaC18H16O9
InChI KeyDXDIHODZARUBLA-DZWQFRRJNA-N
SMILESO.OC(=O)[C@@H](OC(=O)C1=CC=CC=C1)[C@H](OC(=O)C1=CC=CC=C1)C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Melting Point (clear melt)86.0-92.0?C
Assay (Aqueous acid-base Titration)≥98.5 to ≤101.5%
Optical Rotation109.5 ± 0.1? (c=2 in EtOH)
Water Content (Karl Fischer Titration)2.45-7.01% (0.5-1.5 waters)
View more
(+)-Dibenzoyl-D-tartaric acid monohydrate is used as a chiral separation and racemization agent for amines. It is also used as a pharmaceutical raw material.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(+)-Dibenzoyl-D-tartaric acid monohydrate is used as a chiral separation and racemization agent for amines. It is also used as a pharmaceutical raw material.

Solubility
Soluble in methanol. Slightly soluble in water.

Notes
Incompatible with strong oxidizing agents, strong acids and bases.
RUO – Research Use Only

General References:

  1. Chavakula, R.; Mutyala, N. R.; Chennupati, S. R. Industrially Viable Preparation of (R)-3-Quinuclidinol, a Key Building Block of Muscarine M1, M3 Agonists and M3 Antagonists. Org. Prep. Proced. Int. 2013, 45 (6), 507-509.
  2. Gizur, T.; Fogassy, E.; Balint, J.; Egri, G.; Törley, J.; Demeter, A.; Greiner, I. New practical synthesis of Tamsulosin. Chirality 2008, 20 (6), 790-795.