HEPES sodium salt, 99%
HEPES sodium salt, 99%
HEPES sodium salt, 99%
Thermo Scientific Chemicals

HEPES sodium salt, 99%

CAS: 75277-39-3 | C8H17N2NaO4S | 260.28 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Catalog number A16516.14
also known as A16516-14
Price (USD)
50.90
Each
Quantity:
25 g
Request bulk or custom format
Price (USD)
50.90
Each
Chemical Identifiers
CAS75277-39-3
IUPAC Namesodium 2-[4-(2-hydroxyethyl)piperazin-1-yl]ethane-1-sulfonate
Molecular FormulaC8H17N2NaO4S
InChI KeyRDZTWEVXRGYCFV-UHFFFAOYSA-M
SMILES[Na+].OCCN1CCN(CCS([O-])(=O)=O)CC1
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (Titration)≥98.5% (dry wt. basis)
FormCrystals or powder or crystalline powder
Water Content (Karl Fischer Titration)≤3.0%
Appearance (Color)White
HEPES [N-(2-hydroxyethyl)piperazine-N'-(2-ethanesulfonic acid)] is a general purpose buffer for biological research. Its applications include, tissue culture, Phosphorylation and photophosphorylation, fixative Transmission electron microscopy, protein synthesis, preventing binding to non-receptor materials.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
HEPES [N-(2-hydroxyethyl)piperazine-N′-(2-ethanesulfonic acid)] is a general purpose buffer for biological research. Its applications include, tissue culture, Phosphorylation and photophosphorylation, fixative Transmission electron microscopy, protein synthesis, preventing binding to non-receptor materials.

Solubility
It is soluble in water.

Notes
Stable under ordinary conditions. Hygroscopic. Store in cool place. Keep container tightly closed in a dry and well-ventilated place.
RUO – Research Use Only

General References:

  1. H Sands .; LJ Gorey-Feret .; AJ Cocuzza .; FW Hobbs. Biodistribution and metabolism of internally 3H-labeled oligonucleotides. I. Comparison of a phosphodiester and a phosphorothioate. Molecular Pharmacology. 1994, 45, (5), 932-943.
  2. DH Lee .; SY Kim .; JI Hong. A fluorescent pyrophosphate sensor with high selectivity over ATP in water. Angewandte Chemie International Edition. 1994, 43, (36), 4777-4780.