2,5-Difluorobenzylamine, 97%
2,5-Difluorobenzylamine, 97%
2,5-Difluorobenzylamine, 97%
Thermo Scientific Chemicals

2,5-Difluorobenzylamine, 97%

CAS: 85118-06-5 | C7H7F2N | 143.14 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
25 g
Catalog number A16530.06
also known as A16530-06
Price (USD)
88.65
Special offer
Online exclusive
Ends: 30-Jun-2026
104.00
Save 15.35 (15%)
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
88.65
Special offer
Online exclusive
Ends: 30-Jun-2026
104.00
Save 15.35 (15%)
Each
Chemical Identifiers
CAS85118-06-5
IUPAC Name1-(2,5-difluorophenyl)methanamine
Molecular FormulaC7H7F2N
InChI KeyGDFBHCMFIUBEQT-UHFFFAOYSA-N
SMILESNCC1=CC(F)=CC=C1F
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to yellow
FormLiquid
Refractive Index1.4925-1.4975 @ 20?C
Assay (GC)≥96.0%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only