In organic electroluminescent devices, the device with lithium benzoate gives the best performance of all the devices studied. Aromatic multilayers of lithium benzoate were covalently attached to the surface of natural graphite through diazonium chemistry, and the resulting graphite exhibited superior electrochemical performance as anode material of lithium ion batteries.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
In organic electroluminescent devices, the device with lithium benzoate gives the best performance of all the devices studied. Aromatic multilayers of lithium benzoate were covalently attached to the surface of natural graphite through diazonium chemistry, and the resulting graphite exhibited superior electrochemical performance as anode material of lithium ion batteries.
Solubility
Solubility in water is increased by sodium benzoate
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
RUO – Research Use Only
Qinmin Pan,; Hongbo Wang,; Yinghua Jiang. Covalent modification of natural graphite with lithium benzoate multilayers via diazonium chemistry and their application in lithium ion batteries. Electrochemistry Communications. 2007, 9(4), 754-760.
Yasuo Kameda,; Kazuhide Mochiduki,; Masahiro Imano,; Hisashi Naganuma,; Motoya Sasaki,; Yuko Amo,; Takeshi Usuki. Neutron diffraction study of concentrated aqueous lithium benzoate solutions. Journal of Molecular Liquids. 2005, 119 (1-3), 159-166.