1-Aminoindane, 99%
1-Aminoindane, 99%
1-Aminoindane, 99%
Thermo Scientific Chemicals

1-Aminoindane, 99%

CAS: 34698-41-4 | C9H12N | 134.20 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number A16585.03
also known as A16585-03
Price (USD)
40.65
Online Exclusive
44.70
Save 4.05 (9%)
Each
Quantity:
1 g
Request bulk or custom format
Price (USD)
40.65
Online Exclusive
44.70
Save 4.05 (9%)
Each
Chemical Identifiers
CAS34698-41-4
IUPAC Name(1S)-2,3-dihydro-1H-inden-1-aminium
Molecular FormulaC9H12N
InChI KeyXJEVHMGJSYVQBQ-VIFPVBQESA-O
SMILES[NH3+][C@H]1CCC2=CC=CC=C12
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to pale yellow
FormLiquid
Assay (GC)≥97.5%
Refractive Index1.5605-1.5640 @ 20°C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only