2-Chloro-4-nitroaniline, 98+%
2-Chloro-4-nitroaniline, 98+%
2-Chloro-4-nitroaniline, 98+%
Thermo Scientific Chemicals

2-Chloro-4-nitroaniline, 98+%

CAS: 121-87-9 | C6H5ClN2O2 | 172.568 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
1000 g
5000 g
Catalog number A16586.0B
also known as A16586-0B
Price (USD)
148.65
Online Exclusive
165.00
Save 16.35 (10%)
Each
Quantity:
1000 g
Request bulk or custom format
Price (USD)
148.65
Online Exclusive
165.00
Save 16.35 (10%)
Each
Chemical Identifiers
CAS121-87-9
IUPAC Name2-chloro-4-nitroaniline
Molecular FormulaC6H5ClN2O2
InChI KeyLOCWBQIWHWIRGN-UHFFFAOYSA-N
SMILESNC1=CC=C(C=C1Cl)[N+]([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
FormPowder
Appearance (Color)Yellow
Melting Point (clear melt)105.0-111.0?C
Assay (GC)≥98.0%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only