Potassium carbonate, anhydrous, 99%
Potassium carbonate, anhydrous, 99%
Potassium carbonate, anhydrous, 99%
Potassium carbonate, anhydrous, 99%
Thermo Scientific Chemicals

Potassium carbonate, anhydrous, 99%

CAS: 584-08-7 | CK2O3 | 138.21 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10,000 g
500 g
2500 g
Catalog number A16625.0C
also known as A16625-0C
Price (USD)
252.65
Special offer
Online exclusive
Ends: 28-Dec-2026
297.00
Save 44.35
Each
Quantity:
10,000 g
Request bulk or custom format
Price (USD)
252.65
Special offer
Online exclusive
Ends: 28-Dec-2026
297.00
Save 44.35
Each
Chemical Identifiers
CAS584-08-7
IUPAC Namedipotassium carbonate
Molecular FormulaCK2O3
InChI KeyBWHMMNNQKKPAPP-UHFFFAOYSA-L
SMILES[K+].[K+].[O-]C([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Loss on Drying≤1.5% (200?C/constant weight)
Assay (Aqueous acid-base Titration)≥98.5 to ≤101.5%
Identification (FTIR)Conforms
Appearance (Color)White
FormCrystals or powder or crystalline powder or granules
Potassium carbonate has a wide range of industrial and research applications. The major use of potassium carbonate is the manufacture of chemicals, fertilizers, soap and glass. It is used as a drying agent for solvents especially if there are traces of acids present, and as a scavenger of acids produced in organic synthesis. It is used as a buffering agent during the production of wine. It is also used as a fire suppressant in extinguishers. In organic synthesis, it is an extensively used reagent for the alkylation and arylation reactions on carbon, sulfur, nitrogen, and oxygen centres. It is also used in aldol condensation, Michael reaction, acylation, Wittig reaction, Knoevenagel reaction, elimination reaction, epoxide opening, cyclization, metal-catalyzed cross coupling reaction, hydration, and a wide array of reactions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Potassium carbonate has a wide range of industrial and research applications. The major use of potassium carbonate is the manufacture of chemicals, fertilizers, soap and glass. It is used as a drying agent for solvents especially if there are traces of acids present, and as a scavenger of acids produced in organic synthesis. It is used as a buffering agent during the production of wine. It is also used as a fire suppressant in extinguishers. In organic synthesis, it is an extensively used reagent for the alkylation and arylation reactions on carbon, sulfur, nitrogen, and oxygen centres. It is also used in aldol condensation, Michael reaction, acylation, Wittig reaction, Knoevenagel reaction, elimination reaction, epoxide opening, cyclization, metal-catalyzed cross coupling reaction, hydration, and a wide array of reactions.

Solubility
Freely soluble in water. Insoluble in acetone, and alcohol.

Notes
Potassium carbonate is a hygroscopic and deliquescent solid.
RUO – Research Use Only

General References:

  1. In non-aqueous solvents, acts as an economical, moderately-strong base which is widely used in place of other more expensive and hazardous bases in a variety of alkylation and condensation reactions.
  2. For N-alkylation with alkyl halides, see Dibenzyl amine, A11554. For alkylation reactions with dialkyl carbonates, see Dimethyl carbonate, A13104.
  3. Promotes the Wittig olefination reaction in triglyme: Synth. Commun., 22, 513 (1992), or toluene: Synth. Commun., 22, 1691 (1992).
  4. See also Cesium carbonate, 12887.
  5. Mondal, R.; Mallik, A. K. Recent Applications of Potassium Carbonate in Organic Synthesis. Org. Prep. Proced. Int. 2014, 46 (5), 391-434.
  6. Fraley, S. E.; Hall, M. B.; Nennich, T. D. Effect of Variable Water Intake as Mediated by Dietary Potassium Carbonate Supplementation on Rumen dynamics in Lactating Dairy Cows. J. Dairy. Sci. 2015, 98 (5), 3247-3256.
  7. Zhuang, Q. C.; Li, J.; Tian, L. L. Potassium Carbonate as Film Forming Electrolyte Additive for Lithium-ion Batteries. J. Power Sources 2013, 222, 177-183.