1-Cyclohexyl-2-pyrrolidinone, 99%
1-Cyclohexyl-2-pyrrolidinone, 99%
1-Cyclohexyl-2-pyrrolidinone, 99%
Thermo Scientific Chemicals

1-Cyclohexyl-2-pyrrolidinone, 99%

CAS: 6837-24-7 | C10H17NO | 167.252 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
250 g
Catalog number A16638.30
also known as A16638-30
Price (USD)
70.10
Each
Quantity:
250 g
Request bulk or custom format
Price (USD)
70.10
Each
Chemical Identifiers
CAS6837-24-7
IUPAC Name1-cyclohexylpyrrolidin-2-one
Molecular FormulaC10H17NO
InChI KeyPZYDAVFRVJXFHS-UHFFFAOYSA-N
SMILESO=C1CCCN1C1CCCCC1
View more
SpecificationsSpecification SheetSpecification Sheet
Refractive Index1.4975-1.5005 @ 20?C
Appearance (Color)Clear colorless to pale brown
Assay (GC)≥98.5%
FormLiquid
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only