Nonafluoro-1-iodobutane, 97%
Nonafluoro-1-iodobutane, 97%
Nonafluoro-1-iodobutane, 97%
Thermo Scientific Chemicals

Nonafluoro-1-iodobutane, 97%

CAS: 423-39-2 | C4F9I | 345.934 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Catalog number A16671.06
also known as A16671-06
Price (USD)
30.65
Online Exclusive
34.20
Save 3.55 (10%)
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
30.65
Online Exclusive
34.20
Save 3.55 (10%)
Each
Chemical Identifiers
CAS423-39-2
IUPAC Name1,1,1,2,2,3,3,4,4-nonafluoro-4-iodobutane
Molecular FormulaC4F9I
InChI KeyPGRFXXCKHGIFSV-UHFFFAOYSA-N
SMILESFC(F)(F)C(F)(F)C(F)(F)C(F)(F)I
View more
SpecificationsSpecification SheetSpecification Sheet
Identification (FTIR)Conforms
Appearance (Color)Clear colorless or pink to purple
FormLiquid
Assay (GC)≥96.0%
Refractive Index1.3270-1.3330 @ 20°C
Nonafluoro-1-iodobutane is used as iodine atom donor in the synthesis of iodocyclohexane. It is used in agrochemical, pharmaceutical and dyestuff field.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Nonafluoro-1-iodobutane is used as iodine atom donor in the synthesis of iodocyclohexane. It is used in agrochemical, pharmaceutical and dyestuff field.

Solubility
Not miscible in water.

Notes
Light Sensitive. Store in the dark away from oxidizing agents. Incompatible with alkali metals and light.
RUO – Research Use Only