2-Bromo-5-chlorothiophene, 97%
2-Bromo-5-chlorothiophene, 97%
2-Bromo-5-chlorothiophene, 97%
Thermo Scientific Chemicals

2-Bromo-5-chlorothiophene, 97%

CAS: 2873-18-9 | C4H2BrClS | 197.47 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Catalog number A16682.22
also known as A16682-22
Price (USD)
278.65
Online Exclusive
309.00
Save 30.35 (10%)
Each
Quantity:
100 g
Request bulk or custom format
Price (USD)
278.65
Online Exclusive
309.00
Save 30.35 (10%)
Each
Chemical Identifiers
CAS2873-18-9
IUPAC Name2-bromo-5-chlorothiophene
Molecular FormulaC4H2BrClS
InChI KeyZFAJPWYXLYGUJU-UHFFFAOYSA-N
SMILESClC1=CC=C(Br)S1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to pale yellow
Assay (GC)≥96.0%
FormLiquid
Refractive Index1.5930-1.5980 @ 20?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only