2,3-Difluorobenzoic acid, 98%
2,3-Difluorobenzoic acid, 98%
2,3-Difluorobenzoic acid, 98%
Thermo Scientific Chemicals

2,3-Difluorobenzoic acid, 98%

CAS: 4519-39-5 | C7H3F2O2 | 157.10 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number A16717.06
also known as A16717-06
Price (USD)
32.65
Online Exclusive
36.40
Save 3.75 (10%)
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
32.65
Online Exclusive
36.40
Save 3.75 (10%)
Each
Chemical Identifiers
CAS4519-39-5
IUPAC Name2,3-difluorobenzoate
Molecular FormulaC7H3F2O2
InChI KeyJLZVIWSFUPLSOR-UHFFFAOYSA-M
SMILES[O-]C(=O)C1=CC=CC(F)=C1F
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (Aqueous acid-base Titration)≥97.5 to ≤102.5%
FormCrystals or powder or crystalline powder
Assay (Silylated GC)≥97.5%
Appearance (Color)White
Melting Point (clear melt)159.0-166.0?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only