2,3-Difluorophenylacetic acid, 98%
2,3-Difluorophenylacetic acid, 98%
2,3-Difluorophenylacetic acid, 98%
Thermo Scientific Chemicals

2,3-Difluorophenylacetic acid, 98%

CAS: 145689-41-4 | C8H6F2O2 | 172.131 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number A16728.06
also known as A16728-06
Price (USD)
144.65
Special offer
Online exclusive
Ends: 30-Jun-2026
170.00
Save 25.35 (15%)
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
144.65
Special offer
Online exclusive
Ends: 30-Jun-2026
170.00
Save 25.35 (15%)
Each
Chemical Identifiers
CAS145689-41-4
IUPAC Name2-(2,3-difluorophenyl)acetic acid
Molecular FormulaC8H6F2O2
InChI KeyUXSQXUSJGPVOKT-UHFFFAOYSA-N
SMILESOC(=O)CC1=CC=CC(F)=C1F
View more
SpecificationsSpecification SheetSpecification Sheet
FormCrystals or powder or crystalline powder
Assay (HPLC)≥97.5%
Assay (Aqueous acid-base Titration)≥97.5 to ≤102.5%
Melting Point (clear melt)115.0-122.0?C
Appearance (Color)White
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only