4-Methylcyclohexanol, cis + trans, 98%
4-Methylcyclohexanol, cis + trans, 98%
4-Methylcyclohexanol, cis + trans, 98%
Thermo Scientific™

4-Methylcyclohexanol, cis + trans, 98%

CAS: 589-91-3 | C7H14O | 114.19 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 mL
250 mL
1000 mL
Catalog number A16734.AD
also known as A16734-AD
Price (USD)
33.65
Special offer
Online exclusive
Ends: 30-Jun-2026
39.30
Save 5.65 (14%)
Each
Quantity:
50 mL
Request bulk or custom format
Price (USD)
33.65
Special offer
Online exclusive
Ends: 30-Jun-2026
39.30
Save 5.65 (14%)
Each
Chemical Identifiers
CAS589-91-3
IUPAC Name4-methylcyclohexan-1-ol
Molecular FormulaC7H14O
InChI KeyMQWCXKGKQLNYQG-UHFFFAOYSA-N
SMILESCC1CCC(O)CC1
View more
SpecificationsSpecification SheetSpecification Sheet
FormLiquid
Refractive Index1.4565-1.4605 @ 20?C
Appearance (Color)Clear colorless to yellow
Assay (GC)≥97.5%
2-Methylcyclohexanol was used to study the effect of organic solvents on epoxide hydrolase.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Methylcyclohexanol was used to study the effect of organic solvents on epoxide hydrolase.

Solubility
Soluble in water (15 mg/ml at 20°C), ether, alcohols, and many organic solvents.

Notes
Moisture Sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, acid anhydrides, acid chlorides.
RUO – Research Use Only

General References:

  1. Jeanette Lotter et. al.Correlation between the physicochemical properties of organic solvents and their biocompatibility toward epoxide hydrolase activity in whole-cells of a yeast, Rhodotorula sp . Biotechnology Letters. 2004, 26 (15) , 1191-1195.
  2. Halis T. Balaydın et. al.Synthesis and carbonic anhydrase inhibitory properties of novel cyclohexanonyl bromophenol derivatives. Bioorganic & Medicinal Chemistry Letters. 2012, 22(3),1352-1357.