alpha-Toluenesulfonamide, 98%
alpha-Toluenesulfonamide, 98%
alpha-Toluenesulfonamide, 98%
Thermo Scientific Chemicals

alpha-Toluenesulfonamide, 98%

CAS: 4563-33-1 | C7H9NO2S | 171.21 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number A16740.06
also known as A16740-06
Price (USD)
215.65
Online Exclusive
239.00
Save 23.35 (10%)
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
215.65
Online Exclusive
239.00
Save 23.35 (10%)
Each
Chemical Identifiers
CAS4563-33-1
IUPAC Namephenylmethanesulfonamide
Molecular FormulaC7H9NO2S
InChI KeyABOYDMHGKWRPFD-UHFFFAOYSA-N
SMILESNS(=O)(=O)CC1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
FormPowder
Melting Point (clear melt)99.0-105.0?C
Assay (HPLC)≥97.5%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only