4-Fluoro-1-naphthoic acid, 97%
4-Fluoro-1-naphthoic acid, 97%
4-Fluoro-1-naphthoic acid, 97%
Thermo Scientific Chemicals

4-Fluoro-1-naphthoic acid, 97%

CAS: 573-03-5 | C11H7FO2 | 190.173 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number A16778.06
also known as A16778-06
Price (USD)
158.65
Online Exclusive
176.00
Save 17.35 (10%)
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
158.65
Online Exclusive
176.00
Save 17.35 (10%)
Each
Chemical Identifiers
CAS573-03-5
IUPAC Name4-fluoronaphthalene-1-carboxylic acid
Molecular FormulaC11H7FO2
InChI KeyDEWIOKQDRWFLFW-UHFFFAOYSA-N
SMILESOC(=O)C1=C2C=CC=CC2=C(F)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
FormPowder
Appearance (Color)White to cream
Assay (Aqueous acid-base Titration)≥96.0%
Melting Point (clear melt)222.5-228.5?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only