N,N'-Di-n-butylthiourea, 98%
N,N'-Di-n-butylthiourea, 98%
N,N'-Di-n-butylthiourea, 98%
Thermo Scientific Chemicals

N,N'-Di-n-butylthiourea, 98%

CAS: 109-46-6 | C9H20N2S | 188.333 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
250 g
1000 g
Catalog number A16783.0B
also known as A16783-0B
Price (USD)
158.65
Special offer
Online exclusive
Ends: 28-Dec-2026
187.00
Save 28.35
Each
Quantity:
1000 g
Request bulk or custom format
Price (USD)
158.65
Special offer
Online exclusive
Ends: 28-Dec-2026
187.00
Save 28.35
Each
Chemical Identifiers
CAS109-46-6
IUPAC Name1,3-dibutylthiourea
Molecular FormulaC9H20N2S
InChI KeyKFFQABQEJATQAT-UHFFFAOYSA-N
SMILESCCCCNC(=S)NCCCC
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to cream or yellow
FormCrystals or powder or crystalline powder
Assay (HPLC)≥97.5%
Melting Point (clear melt)62.0-68.0?C
N,N'-Di-n-butylthiourea is used in vulcanization accelerator and corrosion inhibitors for metal treatments. It is used as leather, plastic and rubber auxiliary agents.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N,N′-Di-n-butylthiourea is used in vulcanization accelerator and corrosion inhibitors for metal treatments. It is used as leather, plastic and rubber auxiliary agents.

Solubility
Insoluble in water.

Notes
Store in a cool, dry, conditions in well sealed container. Store away from oxidizing agents.
RUO – Research Use Only
Jussi Liippo; Leena Ackermann; Taina Hasan; Arja Laukkanen; Tapio Rantanen; Kaija Lammintausta. Sensitization to thiourea derivatives among Finnish patients with suspected contact dermatitis. Contact Dermatitis.2010, 63 (1), 37-41.