4-Methoxyphenyl isocyanate, 98%
4-Methoxyphenyl isocyanate, 98%
4-Methoxyphenyl isocyanate, 98%
Thermo Scientific Chemicals

4-Methoxyphenyl isocyanate, 98%

CAS: 5416-93-3 | C8H7NO2 | 149.149 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number A16805.06
also known as A16805-06
Price (USD)
46.65
Special offer
Online exclusive
Ends: 28-Dec-2026
55.20
Save 8.55
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
46.65
Special offer
Online exclusive
Ends: 28-Dec-2026
55.20
Save 8.55
Each
Chemical Identifiers
CAS5416-93-3
IUPAC Name1-isocyanato-4-methoxybenzene
Molecular FormulaC8H7NO2
InChI KeyFMDGXCSMDZMDHZ-UHFFFAOYSA-N
SMILESCOC1=CC=C(C=C1)N=C=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
FormLiquid
Assay (GC)≥97.5%
Refractive Index1.5445-1.5490 @ 20?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only