3-(2-Methoxyphenoxy)-1,2-propanediol, 98+%
3-(2-Methoxyphenoxy)-1,2-propanediol, 98+%
3-(2-Methoxyphenoxy)-1,2-propanediol, 98+%
3-(2-Methoxyphenoxy)-1,2-propanediol, 98+%
Thermo Scientific Chemicals

3-(2-Methoxyphenoxy)-1,2-propanediol, 98+%

CAS: 93-14-1 | C10H14O4 | 198.22 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
250 g
1000 g
Catalog number A16827.30
also known as A16827-30
Price (USD)
96.65
Special offer
Online exclusive
Ends: 30-Jun-2026
113.00
Save 16.35 (14%)
Each
Add to cart
Quantity:
250 g
Request bulk or custom format
Price (USD)
96.65
Special offer
Online exclusive
Ends: 30-Jun-2026
113.00
Save 16.35 (14%)
Each
Add to cart
Chemical Identifiers
CAS93-14-1
SpecificationsSpecification SheetSpecification Sheet
Assay (HPLC)≥98.0%
Appearance (Color)White to pale cream
Melting Point (clear melt)77.0-86.0°C
FormPowder or crystalline powder
3-(2-Methoxyphenoxy)-1,2-propanediol is involved in centrally acting muscle relaxant with expectorant properties. It is often used as anesthetic during animal surgery. It is used in the treatment of coughing

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Solubility
Soluble in water (50 mg/ml at 25°C), methanol, DMSO (40 mg/ml at 25°C), DMF, ethanol (50 mg/ml), glycerol and glycol.

Notes
Store in a cool, dry, conditions in well sealed container. Store away from oxidizing agents.
RUO – Research Use Only

General References:

  1. Nesser Ramirez; Matthew Jared; Jeff Hodgden; Wade Toalson. Question: does guaifenesin improve outcomes in patients with cough associated with upper respiratory infection compared with no intervention? The Journal of the Oklahoma State Medical Association.2012, 105 (10), 397-398.
  2. Manujendra Ray; Bela Faltay; Nairmeen Awad Haller. Case report: anaphylactic reaction to guaifenesin. Hospital practice.2009, 37 (1), 60-63.