2,3,4,5,6-Pentafluorostyrene, 98%, stab. with 250ppm 4-tert-butylcatechol
2,3,4,5,6-Pentafluorostyrene, 98%, stab. with 250ppm 4-tert-butylcatechol
2,3,4,5,6-Pentafluorostyrene, 98%, stab. with 250ppm 4-tert-butylcatechol
2,3,4,5,6-Pentafluorostyrene, 98%, stab. with 250ppm 4-tert-butylcatechol
Thermo Scientific Chemicals

2,3,4,5,6-Pentafluorostyrene, 98%, stab. with 250ppm 4-tert-butylcatechol

CAS: 653-34-9 | C8H3F5 | 194.104 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number A16833.14
also known as A16833-14
Price (USD)
252.65
Online Exclusive
281.00
Save 28.35 (10%)
Each
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (USD)
252.65
Online Exclusive
281.00
Save 28.35 (10%)
Each
Add to cart
Chemical Identifiers
CAS653-34-9
IUPAC Name1-ethenyl-2,3,4,5,6-pentafluorobenzene
Molecular FormulaC8H3F5
InChI KeyLVJZCPNIJXVIAT-UHFFFAOYSA-N
SMILESFC1=C(F)C(F)=C(C=C)C(F)=C1F
View more
SpecificationsSpecification SheetSpecification Sheet
Refractive Index1.4440-1.4480 @ 20?C
Assay (GC)≥97.5%
Solution Test10% in diethyl ether: Clear
Solution Test10% in methanol: Clear
Appearance (Color)Clear colorless
View more
2,3,4,5,6-Pentafluorostyrene is used in the synthesis of optically active copolymers having low surface energies by utilizing beta-pinene as another monomer. It is also used in the synthesis of fluorinated and photo crosslinkable liquid prepolymers, which is used for flexible optical waveguides. Polymeric fluorinated nano particles prepared using 2,3,4,5,6-Pentafluorostyrene and styrene is explored in magnetic resonance image (MRI) applications, since it has larger structural design potential compared to traditional systems like emulsions and solutions of smaller molecules.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,3,4,5,6-Pentafluorostyrene is used in the synthesis of optically active copolymers having low surface energies by utilizing beta-pinene as another monomer. It is also used in the synthesis of fluorinated and photo crosslinkable liquid prepolymers, which is used for flexible optical waveguides. Polymeric fluorinated nano particles prepared using 2,3,4,5,6-Pentafluorostyrene and styrene is explored in magnetic resonance image (MRI) applications, since it has larger structural design potential compared to traditional systems like emulsions and solutions of smaller molecules.

Solubility
Immiscible with water.

Notes
Store in a cool and well ventilated area. Keep the container tightly closed, away from sources of ignition and direct sunlight. Incompatible with strong oxidizing agents, strong acids and bases.
RUO – Research Use Only

General References:

  1. Fu, Y.; Lau, Y. T. R.; Weng, L. T.; Ng, K. M.; Chan, C. M. Detection of surface mobility of poly (2, 3, 4, 5, 6-pentafluorostyrene) films by in situ variable-temperature ToF-SIMS and contact angle measurements. J. Colloid Interface Sci. 2014, 431, 180-186.
  2. O'shea, J. P.; Solovyeva, V.; Guo, X.; Zhao, J.; Hadjichristidis, N.; Rodionov, V. O. Sequence-controlled copolymers of 2,3,4,5-pentafluorostyrene: mechanistic insight and application to organocatalysis. Polym. Chem. 2014, 5 (3), 698-701.