2-Ethylphenylhydrazine hydrochloride, 97%
2-Ethylphenylhydrazine hydrochloride, 97%
2-Ethylphenylhydrazine hydrochloride, 97%
Thermo Scientific Chemicals

2-Ethylphenylhydrazine hydrochloride, 97%

CAS: 19398-06-2 | C8H12N2 | 136.20 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
250 g
Catalog number A16853.30
also known as A16853-30
Price (USD)
658.65
Online Exclusive
732.00
Save 73.35 (10%)
Each
Quantity:
250 g
Request bulk or custom format
Price (USD)
658.65
Online Exclusive
732.00
Save 73.35 (10%)
Each
Chemical Identifiers
CAS19398-06-2
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Cream to pale brown
FormCrystals or powder or crystalline powder
Assay (Titration ex Chloride)≥96.0% to ≤104.0%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only