cis-1,2-Bis(diphenylphosphino)ethylene, 97%
cis-1,2-Bis(diphenylphosphino)ethylene, 97%
cis-1,2-Bis(diphenylphosphino)ethylene, 97%
cis-1,2-Bis(diphenylphosphino)ethylene, 97%
Thermo Scientific Chemicals

cis-1,2-Bis(diphenylphosphino)ethylene, 97%

CAS: 983-80-2 | C26H22P2 | 396.41 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number A16859.03
also known as A16859-03
Price (USD)
98.20
Each
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (USD)
98.20
Each
Add to cart
Chemical Identifiers
CAS983-80-2
IUPAC Name[(1Z)-2-(diphenylphosphanyl)ethenyl]diphenylphosphane
Molecular FormulaC26H22P2
InChI KeyNCKJIJSEWKIXAT-DQRAZIAOSA-N
SMILESC(=C\P(C1=CC=CC=C1)C1=CC=CC=C1)\P(C1=CC=CC=C1)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
FormCrystals or crystalline powder
Assay from Supplier's CofA≥96.0%
Phosphorus-31 NMRConforms to cis-isomer structure
Intermediate in organic syntheses.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Intermediate in organic syntheses.

Solubility
Insoluble in water.

Notes
Air Sensitive. Store in a cool, dry, conditions in well sealed container. Store away from oxidizing agents.
RUO – Research Use Only
Chelating ligand.