Naphthalene-2,6-dicarboxylic acid, 98+%
Naphthalene-2,6-dicarboxylic acid, 98+%
Naphthalene-2,6-dicarboxylic acid, 98+%
Thermo Scientific Chemicals

Naphthalene-2,6-dicarboxylic acid, 98+%

CAS: 1141-38-4 | C12H8O4 | 216.192 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Catalog number A16868.14
also known as A16868-14
Price (USD)
209.65
Online Exclusive
233.00
Save 23.35 (10%)
Each
Quantity:
25 g
Request bulk or custom format
Price (USD)
209.65
Online Exclusive
233.00
Save 23.35 (10%)
Each
Chemical Identifiers
CAS1141-38-4
IUPAC Namenaphthalene-2,6-dicarboxylic acid
Molecular FormulaC12H8O4
InChI KeyRXOHFPCZGPKIRD-UHFFFAOYSA-N
SMILESOC(=O)C1=CC2=CC=C(C=C2C=C1)C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to cream
Assay (Silylated GC)≥98.0%
Assay (Aqueous acid-base Titration)≥98.0 to ≤102.0%
FormPowder
Assay (HPLC)≥98.0%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only