3,5-Di-tert-butyl-4-hydroxybenzoic acid, 98%
3,5-Di-tert-butyl-4-hydroxybenzoic acid, 98%
Thermo Scientific Chemicals

3,5-Di-tert-butyl-4-hydroxybenzoic acid, 98%

CAS: 1421-49-4 | C30H42NiO6 | 557.35 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
Catalog number A16880.09
also known as A16880-09
Price (USD)
44.60
Each
Add to cart
Quantity:
10 g
Request bulk or custom format
Price (USD)
44.60
Each
Add to cart
Chemical Identifiers
CAS1421-49-4
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Pale yellow to yellow
Assay (Silylated GC)>97.5%
Assay (Aqueous acid-base Titration)>97.5 to <102.5%
FormPowder
3,5-Di-tert-butyl-4-hydroxybenzoic acid was used in the synthesis of antistress agents. It is used as pharmaceutical intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3,5-Di-tert-butyl-4-hydroxybenzoic acid was used in the synthesis of antistress agents. It is used as pharmaceutical intermediates.

Solubility
Insoluble in water.

Notes
Store in a cool, dry, conditions in well sealed container. Store away from oxidizing agents.
RUO – Research Use Only
Karyophyllis C Tsiakitzis; Eleni A Rekka; Angeliki P Kourounakis; Panos N Kourounakis. Novel compounds designed as antistress agents. Journal of medicinal and pharmaceutical chemistry.2009, 52 (22), 7315-7318.