Employed as a pharmaceutical intermediate. It is also used in organic synthesis.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Employed as a pharmaceutical intermediate. It is also used in organic synthesis.
Solubility
Very faint turbidity in methanol.
Notes
Store away from oxidizing agents. Keep the container tightly sealed and store in cool, dry condition in well ventilated place.
RUO – Research Use Only
General References:
- Sunkyung Lee.; Stewart P. Frescas & David E. Nichols. A New, Simple Procedure for the Preparation of 8-Methoxy-2-Tetralone.An International Journal for Rapid Communication of Synthetic Organic Chemistry,1995, 25(18), 2775-2780.
- Gang Lu.; Robert Franzén.; Qian Zhang.; Youjun Xu. Palladium charcoal-catalyzed, ligandless Suzuki reaction by using tetraarylborates in water.Tetrahedron Lett.2005, 46(24), 4255-4259.