2-Fluorotoluene, 99+%
2-Fluorotoluene, 99+%
2-Fluorotoluene, 99+%
2-Fluorotoluene, 99+%
Thermo Scientific Chemicals

2-Fluorotoluene, 99+%

CAS: 95-52-3 | C7H7F | 110.131 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
2500 g
Catalog number A16912.0E
also known as A16912-0E
Price (USD)
362.65
Online Exclusive
403.00
Save 40.35 (10%)
Each
Add to cart
Quantity:
2500 g
Request bulk or custom format
Price (USD)
362.65
Online Exclusive
403.00
Save 40.35 (10%)
Each
Add to cart
Chemical Identifiers
CAS95-52-3
IUPAC Name1-fluoro-2-methylbenzene
Molecular FormulaC7H7F
InChI KeyMMZYCBHLNZVROM-UHFFFAOYSA-N
SMILESCC1=CC=CC=C1F
View more
SpecificationsSpecification SheetSpecification Sheet
Identification (FTIR)Conforms
FormLiquid
Refractive Index1.4725-1.4745 @ 20?C
Appearance (Color)Clear colorless
Assay (GC)≥99.0%
It is employed in organic synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is employed in organic synthesis.

Solubility
Not miscible or difficult to mix in water.

Notes
Store away from oxidizing agents. Keep the container tightly sealed and store in cool, dry condition in well ventilated place.
RUO – Research Use Only

General References:

  1. Ted Schaefer.; Rudy Sebastian.; Richard P. Veregin.; Reino Laatikainen. Some unique properties of in 2-fluorotoluene derivatives. Conformational dependence.Can. J. Chem.,1983,61(1), 29-36.
  2. Sushil K. Maurya.; M. K. Gurjar.; K. M. Malshe.; P. T. Patil.; M. K. Dongare.; and Erhard Kemnitz. Solid acid catalysts for fluorotoluene nitration using nitric acid.Green Chem.,2003,5720-723.