(R)-(+)-1-(1-Naphthyl)ethylamine, 99%
(R)-(+)-1-(1-Naphthyl)ethylamine, 99%
(R)-(+)-1-(1-Naphthyl)ethylamine, 99%
Thermo Scientific Chemicals

(R)-(+)-1-(1-Naphthyl)ethylamine, 99%

CAS: 3886-70-2 | C12H13N | 171.243 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number A16918.06
also known as A16918-06
Price (USD)
137.65
Online Exclusive
153.00
Save 15.35 (10%)
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
137.65
Online Exclusive
153.00
Save 15.35 (10%)
Each
Chemical Identifiers
CAS3886-70-2
IUPAC Name(1R)-1-(naphthalen-1-yl)ethan-1-aminium
Molecular FormulaC12H14N
InChI KeyRTCUCQWIICFPOD-SECBINFHSA-O
SMILESC[C@@H]([NH3+])C1=C2C=CC=CC2=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to yellow
FormLiquid
Assay (GC)≥96.0%
CommentMay darken on storage
Optical Rotation+80° to +82° (neat)
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only