4-Isopropylanisole, 95%
4-Isopropylanisole, 95%
4-Isopropylanisole, 95%
Thermo Scientific Chemicals

4-Isopropylanisole, 95%

CAS: 4132-48-3 | C10H14O | 150.22 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Catalog number A16970.14
also known as A16970-14
Price (USD)
144.65
Online exclusive
161.00
Save 16.35
Each
Quantity:
25 g
Request bulk or custom format
Price (USD)
144.65
Online exclusive
161.00
Save 16.35
Each
Chemical Identifiers
CAS4132-48-3
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to yellow
Assay (GC)≥94.0%
FormLiquid
Refractive Index1.5010-1.5070 @ 20?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only