2,3-Difluorophenol, 98+%
2,3-Difluorophenol, 98+%
2,3-Difluorophenol, 98+%
Thermo Scientific Chemicals

2,3-Difluorophenol, 98+%

CAS: 6418-38-8 | C6H4F2O | 130.094 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number A16971.06
also known as A16971-06
Price (USD)
55.65
Special offer
Online exclusive
Ends: 30-Jun-2026
65.60
Save 9.95 (15%)
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
55.65
Special offer
Online exclusive
Ends: 30-Jun-2026
65.60
Save 9.95 (15%)
Each
Chemical Identifiers
CAS6418-38-8
IUPAC Name2,3-difluorophenol
Molecular FormulaC6H4F2O
InChI KeyRPEPGIOVXBBUMJ-UHFFFAOYSA-N
SMILESOC1=CC=CC(F)=C1F
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (GC)≥98.0%
FormFused solid or clear liquid as melt
Appearance (Color)Colorless to white to cream
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only