2-Ethylbutyric acid, 98%
2-Ethylbutyric acid, 98%
2-Ethylbutyric acid, 98%
Thermo Scientific Chemicals

2-Ethylbutyric acid, 98%

CAS: 88-09-5 | C6H12O2 | 116.16 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mL
1000 mL
Catalog number A16989.AK
also known as A16989-AK
Price (USD)
50.65
Online Exclusive
56.60
Save 5.95 (11%)
Each
Quantity:
250 mL
Request bulk or custom format
Price (USD)
50.65
Online Exclusive
56.60
Save 5.95 (11%)
Each
Chemical Identifiers
CAS88-09-5
IUPAC Name2-ethylbutanoic acid
Molecular FormulaC6H12O2
InChI KeyOXQGTIUCKGYOAA-UHFFFAOYSA-N
SMILESCCC(CC)C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to pale yellow
FormLiquid
Assay (Aqueous acid-base Titration)≥97.5 to ≤102.5%
Assay (Silylated GC)≥97.5%
Identification (FTIR)Conforms
View more
Diethyl acetic acid (88-09-5) is used in flavoring agent and as an intermediate for pharmaceuticals and dyestuffs. It is also employed in the manufacture of esters and other chemicals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Diethyl acetic acid (88-09-5) is used in flavoring agent and as an intermediate for pharmaceuticals and dyestuffs. It is also employed in the manufacture of esters and other chemicals.

Solubility
Sparingly soluble with the usual organic solvents. Solubility in water 18 g/L (20°C).

Notes
Store away from oxidizing agents. Keep the container tightly sealed and store in cool, dry condition in well ventilated place.
RUO – Research Use Only

General References:

  1. P.E. Keane.; J. Simiand.; E. Mendes.; V. Santucci.; M. Morre. The effects of analogues of valproic acid on seizures induced by pentylenetetrazol and GABA content in brain of mice.Neuropharmacology,1983,22(7), 875-879.
  2. Hiroshi Miwa.; Magobei Yamamoto. Liquid chromatographic determination of free and total fatty acids in milk and milk products as their 2-nitrophenylhydrazides.J. Chromatogr. A,1990,523,235-246.