Sodium hydrogen carbonate, 99%
Sodium hydrogen carbonate, 99%
Sodium hydrogen carbonate, 99%
Thermo Scientific Chemicals

Sodium hydrogen carbonate, 99%

CAS: 144-55-8 | CHNaO3 | 84.01 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10,000 g
500 g
2500 g
Catalog number A17005.0C
also known as A17005-0C
Price (USD)
209.65
Special offer
Online exclusive
Ends: 30-Jun-2026
246.00
Save 36.35 (15%)
Each
Quantity:
10,000 g
Request bulk or custom format
Price (USD)
209.65
Special offer
Online exclusive
Ends: 30-Jun-2026
246.00
Save 36.35 (15%)
Each
Chemical Identifiers
CAS144-55-8
IUPAC Namesodium hydrogen carbonate
Molecular FormulaCHNaO3
InChI KeyUIIMBOGNXHQVGW-UHFFFAOYSA-M
SMILES[Na+].OC([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (Aqueous acid-base Titration)≥98.5 to ≤101.5%
FormPowder
Appearance (Color)White
Identification (FTIR)Conforms
Sodium hydrogen carbonate is used as a laboratory chemical, a leavening agent, a neutralizer, as an active component in fire extinguishers, as a source of carbon dioxide and pH balancer in pools, as a cattle feed supplement and in spas and garden ponds. It is utilized for paint and corrosion removal, baking powder, as a mild abrasive and in cleaning compounds. In the field of medicine, it is used as an antacid to treat acid indigestion and heartburn.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Sodium hydrogen carbonate is used as a laboratory chemical, a leavening agent, a neutralizer, as an active component in fire extinguishers, as a source of carbon dioxide and pH balancer in pools, as a cattle feed supplement and in spas and garden ponds. It is utilized for paint and corrosion removal, baking powder, as a mild abrasive and in cleaning compounds. In the field of medicine, it is used as an antacid to treat acid indigestion and heartburn.

Solubility
Soluble in water and methanol. Insoluble in ethanol.

Notes
Incompatible with strong acids and strong oxidizing agents.
RUO – Research Use Only

General References:

  1. Yang, X.; Wu, J.; Mao, X.; Jamison, T. F.; Hatton, T. A. Microwave assisted synthesis of cyclic carbonates from olefins with sodium bicarbonates as the C1 source. Chem. Commun. 2014, 50 (24), 3245-3248.
  2. Maurya, M. R.; Kumar, N. Sodium bicarbonate assisted oxidation, by H2O2, of styrene and cyclohexene using polymer grafted dioxidomolybdenum(VI) complex as a catalyst. J. Mol. Catal. A: Chem. 2015, 406, 204-212.