3,4-Dichlorobenzoyl chloride, 97%
3,4-Dichlorobenzoyl chloride, 97%
3,4-Dichlorobenzoyl chloride, 97%
3,4-Dichlorobenzoyl chloride, 97%
Thermo Scientific Chemicals

3,4-Dichlorobenzoyl chloride, 97%

CAS: 3024-72-4 | C7H3Cl3O | 209.45 g/mol
Have Questions?
Catalog NumberQuantity
A17006.22
also known as A17006-22
100 g
Catalog number A17006.22
also known as A17006-22
Price (USD)
107.65
Special offer
Online exclusive
Ends: 31-Dec-2025
126.00
Save 18.35 (15%)
Each
Add to cart
Quantity:
100 g
Request bulk or custom format
Price (USD)
107.65
Special offer
Online exclusive
Ends: 31-Dec-2025
126.00
Save 18.35 (15%)
Each
Add to cart
Chemical Identifiers
CAS3024-72-4
IUPAC Name3,4-dichlorobenzoyl chloride
Molecular FormulaC7H3Cl3O
InChI KeyVTXNOVCTHUBABW-UHFFFAOYSA-N
SMILESClC(=O)C1=CC=C(Cl)C(Cl)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to yellow or colorless to yellow
Melting Point (clear melt)28.5-37.5?C
FormFused solid or liquid as melt
Assay (GC)≥96.0%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only