3-(Trifluoromethyl)benzonitrile, 98%
3-(Trifluoromethyl)benzonitrile, 98%
3-(Trifluoromethyl)benzonitrile, 98%
Thermo Scientific Chemicals

3-(Trifluoromethyl)benzonitrile, 98%

CAS: 368-77-4 | C8H4F3N | 171.122 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Catalog number A17029.22
also known as A17029-22
Price (USD)
165.65
Online Exclusive
184.00
Save 18.35 (10%)
Each
Quantity:
100 g
Request bulk or custom format
Price (USD)
165.65
Online Exclusive
184.00
Save 18.35 (10%)
Each
Chemical Identifiers
CAS368-77-4
IUPAC Name3-(trifluoromethyl)benzonitrile
Molecular FormulaC8H4F3N
InChI KeyOGOBINRVCUWLGN-UHFFFAOYSA-N
SMILESFC(F)(F)C1=CC=CC(=C1)C#N
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to yellow to orange
FormLiquid as melt
Refractive Index1.4575-1.4615 as melt
Assay (GC)≥97.5%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only