Ethyl 2-thiouracil-5-carboxylate, 99%
Ethyl 2-thiouracil-5-carboxylate, 99%
Ethyl 2-thiouracil-5-carboxylate, 99%
Thermo Scientific Chemicals

Ethyl 2-thiouracil-5-carboxylate, 99%

CAS: 38026-46-9 | C7H8N2O3S | 200.21 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number A17046.06
also known as A17046-06
Price (USD)
31.65
Online Exclusive
35.40
Save 3.75 (11%)
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
31.65
Online Exclusive
35.40
Save 3.75 (11%)
Each
Chemical Identifiers
CAS38026-46-9
IUPAC Nameethyl 4-oxo-2-sulfanylidene-1,2,3,4-tetrahydropyrimidine-5-carboxylate
Molecular FormulaC7H8N2O3S
InChI KeyFQFSHLBWRUOCPX-UHFFFAOYSA-N
SMILESCCOC(=O)C1=CNC(=S)NC1=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
FormPowder
Assay (Aqueous acid-base Titration)≥98.5 to ≤101.5%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only