2,4-Difluorobenzaldehyde, 98%
2,4-Difluorobenzaldehyde, 98%
2,4-Difluorobenzaldehyde, 98%
Thermo Scientific Chemicals

2,4-Difluorobenzaldehyde, 98%

CAS: 1550-35-2 | C7H4F2O | 142.11 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number A17052.14
also known as A17052-14
Price (USD)
160.65
Special offer
Online exclusive
Ends: 30-Jun-2026
189.00
Save 28.35 (15%)
Each
Quantity:
25 g
Request bulk or custom format
Price (USD)
160.65
Special offer
Online exclusive
Ends: 30-Jun-2026
189.00
Save 28.35 (15%)
Each
Chemical Identifiers
CAS1550-35-2
IUPAC Name2,4-difluorobenzaldehyde
Molecular FormulaC7H4F2O
InChI KeyWCGPCBACLBHDCI-UHFFFAOYSA-N
SMILESFC1=CC=C(C=O)C(F)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to yellow
Assay (GC)≥97.5%
Free acid (titration)≤1%
Refractive Index1.4970-1.5010 @ 20?C
FormLiquid
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only