Sodium salicylate, 99%
Sodium salicylate, 99%
Sodium salicylate, 99%
Thermo Scientific Chemicals

Sodium salicylate, 99%

CAS: 54-21-7 | C7H5NaO3 | 160.104 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
1000 g
Catalog number A17056.30
also known as A17056-30
Price (USD)
59.10
Each
Quantity:
250 g
Request bulk or custom format
Price (USD)
59.10
Each
Chemical Identifiers
CAS54-21-7
IUPAC Namesodium 2-hydroxybenzoate
Molecular FormulaC7H5NaO3
InChI KeyABBQHOQBGMUPJH-UHFFFAOYSA-M
SMILES[Na+].OC1=CC=CC=C1C([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White or colorless to pale cream
FormCrystals or powder or crystalline powder
Assay (Non-aqueous acid-base Titration)≥98.5 to ≤101.5% (UK Sourced Material)
Assay from Supplier's CofA≥98.5% (US Sourced Material)
CommentMaterial Sourced in UK and US
View more
Sodium salicylate is used as an analgesic and antipyretic. It also acts as a non-steroidal anti-inflammatory drug (NSAID). Further, it is used as a phosphor for the detection of vacuum ultraviolet radiation and electrons. It serves as a neurofibromatosis kappa B inhibitor and reducer of oxidative stress. It finds applications as a drug with the function of being antifebrile and anti-rheumatic.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Sodium salicylate is used as an analgesic and antipyretic. It also acts as a non-steroidal anti-inflammatory drug (NSAID). Further, it is used as a phosphor for the detection of vacuum ultraviolet radiation and electrons. It serves as a neurofibromatosis kappa B inhibitor and reducer of oxidative stress. It finds applications as a drug with the function of being antifebrile and anti-rheumatic.

Solubility
Soluble in water, 1,4-dioxane, ethanol and glycerol.

Notes
Light sensitive. Store in a cool place. Incompatible with strong oxidizing agents, strong acidsmineral acids, metallic salts and iodine.
RUO – Research Use Only

General References:

  1. Younatham, R.; Siefers, K.; Wegman-Points, L.; Pierce, G. Sodium salicylate prevents TNF alpha-induced reductions in insulin-stimulated eNOSser1177 phosphorylation but not Akt in human aortic endothelial cells. FASEB J. 2015, 29 (1), 802-805.
  2. Li, Q.; Qi, Y.; Gao, C. Chemical regeneration of spent powdered activated carbon used in decolorization of sodium salicylate for the pharmaceutical industry. J. Cleaner Prod. 2015, 86, 424-431.