2,2-Dimethylbutyric acid, 97%
2,2-Dimethylbutyric acid, 97%
2,2-Dimethylbutyric acid, 97%
Thermo Scientific Chemicals

2,2-Dimethylbutyric acid, 97%

CAS: 595-37-9 | C6H12O2 | 116.16 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
250 g
Catalog number A17072.30
also known as A17072-30
Price (USD)
266.65
Online Exclusive
296.00
Save 29.35 (10%)
Each
Quantity:
250 g
Request bulk or custom format
Price (USD)
266.65
Online Exclusive
296.00
Save 29.35 (10%)
Each
Chemical Identifiers
CAS595-37-9
IUPAC Name2,2-dimethylbutanoic acid
Molecular FormulaC6H12O2
InChI KeyVUAXHMVRKOTJKP-UHFFFAOYSA-N
SMILESCCC(C)(C)C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to pale yellow
FormLiquid
Assay (Aqueous acid-base Titration)≥96.0 to ≤104.0%
Assay (Silylated GC)≥96.0%
Refractive Index1.4125-1.4175 @ 20?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only